4-Fluor-3-(trifluormethoxy)benzylbromid 98%
Lieferant: Apollo Scientific
|
Gefahr
|
PC49274-1G
PC49274-5G
PC49274-10G
APOSPC49274-1GEA
226
EUR
APOSPC49274-1G
APOSPC49274-5G
APOSPC49274-10G
4-Fluor-3-(trifluormethoxy)benzylbromid 98%
4-Fluor-3-(trifluormethoxy)benzylbromid
Formel:
C₈H₅BrF₄O Molecular Weight: 273.02 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06660256 CAS-Nummer: 86256-50-0 UN: 3265 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...