3-Hydroxy-5-(trifluoromethyl)benzaldehyde

Lieferant: Apollo Scientific
Achtung

PC49281-1G PC49281-250MG PC49281-5G
APOSPC49281-1GEA 145 EUR
APOSPC49281-1G APOSPC49281-250MG APOSPC49281-5G
3-Hydroxy-5-(trifluoromethyl)benzaldehyde
3-Hydroxy-5-(trifluoromethyl)benzaldehyde
Formel: C₈H₅F₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD16999720
CAS-Nummer: 1243312-51-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR