1,1,2,3,3-Pentafluoropropane 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: HFC-245ea

PC49289-1G
APOSPC49289-1GEA 232 EUR
APOSPC49289-1G
1,1,2,3,3-Pentafluoropropane 97%
1,1,2,3,3-Pentafluoropropane
Formel: C₃H₃F₅
Molecular Weight: 134,048 g/mol
Siedepunkt: 39…40 °C (748 mm Hg)
Lagertemperatur: Raumtemperatur
MDL: MFCD22566640
CAS-Nummer: 24270-66-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR