2-Bromo-4-(trifluoromethoxy)benzeneboronic acid

Lieferant: Apollo Scientific
Achtung

PC49295-1G PC49295-250MG PC49295-5G
APOSPC49295-1GEA 0 EUR
APOSPC49295-1G APOSPC49295-250MG APOSPC49295-5G
2-Bromo-4-(trifluoromethoxy)benzeneboronic acid
2-Bromo-4-(trifluoromethoxy)benzeneboronic acid
Formel: C₇H₅BBrF₃O₃
Lagertemperatur: Kühlschrank
MDL: MFCD19237163
CAS-Nummer: 959997-86-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR