3-Brom-5-fluoranilin 97%

Lieferant: Apollo Scientific
Achtung

PC49300-1G PC49300-25G PC49300-5G
APOSPC49300-1GEA 40.6 EUR
APOSPC49300-1G APOSPC49300-25G APOSPC49300-5G
3-Brom-5-fluoranilin 97%
3-Brom-5-fluoranilin
Formel: C₆H₅BrFN
Lagertemperatur: Raumtemperatur
MDL: MFCD07779529
CAS-Nummer: 134168-97-1
UN: 2810
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR