3-Brom-5-fluoranilin 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC49300-1G
PC49300-25G
PC49300-5G
APOSPC49300-1GEA
40.6
EUR
APOSPC49300-1G
APOSPC49300-25G
APOSPC49300-5G
3-Brom-5-fluoranilin 97%
3-Brom-5-fluoranilin
Formel:
C₆H₅BrFN Lagertemperatur: Raumtemperatur |
MDL:
MFCD07779529 CAS-Nummer: 134168-97-1 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...