5-(Pentafluorothio)-1,3-benzoxazole

Lieferant: Apollo Scientific
Achtung

PC49305-1G PC49305-250MG PC49305-5G
APOSPC49305-1GEA 0 EUR
APOSPC49305-1G APOSPC49305-250MG APOSPC49305-5G
5-(Pentafluorothio)-1,3-benzoxazole
5-(Pentafluorothio)-1,3-benzoxazole
Formel: C₇H₄F₅NOS
Lagertemperatur: Raumtemperatur
MDL: MFCD22370119
CAS-Nummer: 1379812-05-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR