5-(Pentafluorothio)-1H-benzimidazole

Lieferant: Apollo Scientific
Achtung

Synonyme: (1H-Benzimidazol-5-yl)sulphur pentafluoride, 5-(Pentafluorosulphanyl)-1H-benzimidazole

PC49306-1G PC49306-250MG
APOSPC49306-1GEA 279 EUR
APOSPC49306-1G APOSPC49306-250MG
5-(Pentafluorothio)-1H-benzimidazole
5-(Pentafluorothio)-1H-benzimidazole
Formel: C₇H₅F₅N₂S
Molecular Weight: 244.185 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22370117
CAS-Nummer: 663179-60-0
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR