2-(Trifluoromethyl)isonicotinonitrile ≥95%

Lieferant: Apollo Scientific
Achtung

PC49309-250MG PC49309-1G
APOSPC49309-250MGEA 0 EUR
APOSPC49309-250MG APOSPC49309-1G
2-(Trifluoromethyl)isonicotinonitrile ≥95%
2-(Trifluoromethyl)isonicotinonitrile
Formel: C₇H₃F₃N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11848100
CAS-Nummer: 916210-02-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR