N-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-3-(hydrazinocarbonyl)-5-methyl-4-isoxazolecarboxamide 97%

Lieferant: Apollo Scientific

PC4931-500MG
APOSPC4931-500MGEA 976 EUR
APOSPC4931-500MG
N-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-3-(hydrazinocarbonyl)-5-methyl-4-isoxazolecarboxamide 97%
N-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-3-(hydrazinocarbonyl)-5-methyl-4-isoxazolecarboxamide
Formel: C₁₂H₉ClF₃N₅O₃
Molecular Weight: 363,69 g/mol
Schmelzpunkt: 146…148 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00664740
CAS-Nummer: 338397-37-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR