1,4-Diethynyl-2-fluorobenzene
Lieferant: Apollo Scientific
|
Achtung
|
PC49311-1G
PC49311-5G
PC49311-250MG
APOSPC49311-1GEA
58
EUR
APOSPC49311-1G
APOSPC49311-5G
APOSPC49311-250MG
1,4-Diethynyl-2-fluorobenzene
1,4-Diethynyl-2-fluorobenzene
Formel:
C₁₀H₅F Schmelzpunkt: 86…89 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD22580725 CAS-Nummer: 137000-66-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...