2-(Trifluormethyl)-4-pyridinol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Hydroxy-2-(trifluoromethyl)pyridine
PC49312-1G
APOSPC49312-1GEA
21.9
EUR
APOSPC49312-1G
2-(Trifluormethyl)-4-pyridinol
2-(Trifluoromethyl)pyridin-4-ol
Formel:
C₆H₄F₃NO Molecular Weight: 163,10 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01571202 CAS-Nummer: 170886-13-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...