2-(Trifluoromethyl)isonicotinaldehyde

Lieferant: Apollo Scientific
Achtung

PC49315-1G PC49315-250MG
APOSPC49315-1GEA 270 EUR
APOSPC49315-1G APOSPC49315-250MG
2-(Trifluoromethyl)isonicotinaldehyde
2-(Trifluoromethyl)isonicotinaldehyde
Formel: C₇H₄F₃NO
Lagertemperatur: Kühlschrank
MDL: MFCD06410678
CAS-Nummer: 108338-20-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR