6-Nitro-2-(pentafluoroethyl)-1H-benzimidazole
Lieferant: Apollo Scientific
Synonyme:
6-Nitro-2-(perfluoroethyl)-1H-benzimidazole
PC49317-1G
PC49317-250MG
PC49317-5G
APOSPC49317-1GEA
161
EUR
APOSPC49317-1G
APOSPC49317-250MG
APOSPC49317-5G
6-Nitro-2-(pentafluoroethyl)-1H-benzimidazole
6-Nitro-2-(pentafluoroethyl)-1H-benzimidazole
Formel:
C₉H₄F₅N₃O₂ Schmelzpunkt: 140…141 °C Flammpunkt: >110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD18074389 CAS-Nummer: 4229-01-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...