2-{2-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]hydrazono}pentanedioic acid 97%

Lieferant: Apollo Scientific

PC4932-1G
APOSPC4932-1GEA 576 EUR
APOSPC4932-1G
2-{2-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]hydrazono}pentanedioic acid 97%
2-{2-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]hydrazono}pentanedioic acid
Formel: C₁₁H₉ClF₃N₃O₄
Molecular Weight: 339,66 g/mol
Schmelzpunkt: 222…224 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00173606
CAS-Nummer: 338422-72-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR