3'-Bromo-2,2,2-trifluoro-5'-(trifluoromethyl)acetophenone 95%

Lieferant: Apollo Scientific
Achtung

PC49322-1G PC49322-250MG
APOSPC49322-1GEA 290 EUR
APOSPC49322-1G APOSPC49322-250MG
3'-Bromo-2,2,2-trifluoro-5'-(trifluoromethyl)acetophenone 95%
3'-Bromo-2,2,2-trifluoro-5'-(trifluoromethyl)acetophenone
Formel: C₉H₃BrF₆O
Lagertemperatur: Raumtemperatur
MDL: MFCD23098942
CAS-Nummer: 1132701-00-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR