3-Bromo-2-fluorophenacyl bromide 95%

Lieferant: Apollo Scientific
Gefahr

PC49325-1G PC49325-250MG PC49325-5G
APOSPC49325-1GEA 0 EUR
APOSPC49325-1G APOSPC49325-250MG APOSPC49325-5G
3-Bromo-2-fluorophenacyl bromide 95%
3-Bromo-2-fluorophenacyl bromide
Formel: C₈H₅Br₂FO
Lagertemperatur: Raumtemperatur
MDL: MFCD16140216
CAS-Nummer: 1204333-47-0
UN: 3261
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR