4-Chlor-3-(trifluormethoxy)benzylbromid 97%
Lieferant: Apollo Scientific
|
Gefahr
|
PC49329-1G
PC49329-5G
PC49329-25G
APOSPC49329-1GEA
0
EUR
APOSPC49329-1G
APOSPC49329-5G
APOSPC49329-25G
4-Chlor-3-(trifluormethoxy)benzylbromid 97%
4-Chlor-3-(trifluormethoxy)benzylbromid
Formel:
C₈H₅BrClF₃O Lagertemperatur: Raumtemperatur |
MDL:
MFCD06660249 CAS-Nummer: 886500-93-2 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...