2-Methoxy-5-(trifluormethoxy)benzylbromid 98%

Lieferant: Apollo Scientific
Gefahr

PC49330-10G PC49330-5G PC49330-1G
APOSPC49330-10GEA 0 EUR
APOSPC49330-10G APOSPC49330-5G APOSPC49330-1G
2-Methoxy-5-(trifluormethoxy)benzylbromid 98%
2-Methoxy-5-(trifluormethoxy)benzylbromid
Formel: C₉H₈BrF₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD04115961
CAS-Nummer: 478484-53-6
UN: 3265
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR