Ethyl 2,2-Difluoro-2-(trimethylsilyl)acetate 95%

Lieferant: Apollo Scientific
Gefahr

Synonyme: Ethyl 2,2-difluoro-2-(trimethylsilyl)acetate, 2,2-Difluoro-2-(trimethylsilyl)acetic acid Ethyl ester

PC49333-25G PC49333-1G PC49333-5G
APOSPC49333-25GEA 435 EUR
APOSPC49333-25G APOSPC49333-1G APOSPC49333-5G
Ethyl 2,2-Difluoro-2-(trimethylsilyl)acetate 95%
Ethyl 2,2-Difluoro-2-(trimethylsilyl)acetate
Formel: C₇H₁₄F₂O₂Si
Siedepunkt: 55 °C/20mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD04973092
CAS-Nummer: 205865-67-4
UN: 1993
ADR: 3,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR