3-Methoxy-5-(trifluormethyl)phenylacetonitril 98%

Lieferant: Apollo Scientific

PC49341-1G PC49341-5G PC49341-10G
APOSPC49341-1GEA 0 EUR
APOSPC49341-1G APOSPC49341-5G APOSPC49341-10G
3-Methoxy-5-(trifluormethyl)phenylacetonitril 98%
3-Methoxy-5-(trifluormethyl)phenylacetonitril
Formel: C₁₀H₈F₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD09025415
CAS-Nummer: 916421-02-8
UN: 3276
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR