Difluoro(pyridin-2-yl)acetic acid hydrochloride

Lieferant: Apollo Scientific
Achtung

PC49343-1G PC49343-5G
APOSPC49343-1GEA 0 EUR
APOSPC49343-1G APOSPC49343-5G
Difluoro(pyridin-2-yl)acetic acid hydrochloride
Difluoro(pyridin-2-yl)acetic acid hydrochloride
Formel: C₇H₅F₂NO₂·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD22392210
CAS-Nummer: 1421603-45-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR