2,4-Difluoro-3-(trifluoromethyl)styrene

Lieferant: Apollo Scientific
Achtung

PC49345-1G PC49345-250MG
APOSPC49345-1GEA 0 EUR
APOSPC49345-1G APOSPC49345-250MG
2,4-Difluoro-3-(trifluoromethyl)styrene
2,4-Difluoro-3-(trifluoromethyl)styrene
Formel: C₉H₅F₅
Lagertemperatur: Kühlschrank
MDL: MFCD24369700
CAS-Nummer: 1432075-80-3
UN: 1993
ADR: 3,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR