4-Bromo-3-(trifluoromethyl)phenylacetylene 97%

Lieferant: Apollo Scientific
Achtung

PC49346-1G PC49346-250MG
APOSPC49346-1GEA 0 EUR
APOSPC49346-1G APOSPC49346-250MG
4-Bromo-3-(trifluoromethyl)phenylacetylene 97%
4-Bromo-3-(trifluoromethyl)phenylacetylene
Formel: C₉H₄BrF₃
Schmelzpunkt: 50 °C
Lagertemperatur: Kühlschrank
MDL: MFCD24369704
CAS-Nummer: 142476-42-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR