1-(3-Chlor-5-(trifluormethyl)-2-pyridinyl)-1H-indol-3-carbaldehyd 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC4935-1G
PC4935-500MG
APOSPC4935-1GEA
434
EUR
APOSPC4935-1G
APOSPC4935-500MG
1-(3-Chlor-5-(trifluormethyl)-2-pyridinyl)-1H-indol-3-carbaldehyd 97%
1-(3-Chlor-5-(trifluormethyl)-2-pyridinyl)-1H-indol-3-carbaldehyd
Formel:
C₁₅H₈ClF₃N₂O Schmelzpunkt: 132…134 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00664496 CAS-Nummer: 338770-32-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...