(1R,2S)-2-Fluorocyclopropanamine-4-methylbenzenesulphonate 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: (1R,2S)-2-Fluorocyclopropanamine 4-methylbenzenesulphonate

PC49351-5G
APOSPC49351-5GEA 59.6 EUR
APOSPC49351-5G
(1R,2S)-2-Fluorocyclopropanamine-4-methylbenzenesulphonate 98%
(1R,2S)-2-Fluorocyclopropanamine-4-methylbenzenesulphonate
Formel: C₁₀H₁₄FNO₃S
Molecular Weight: 247.29 g/mol
Schmelzpunkt: 178…179 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01861147
CAS-Nummer: 143062-84-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR