2-[3-Chloro-5-(trifluoromethyl)pyridin-2-ylaminoethyl]amino-5-chloroaniline

Lieferant: Apollo Scientific

PC4936-500MG
APOSPC4936-500MGEA 855 EUR
APOSPC4936-500MG
2-[3-Chloro-5-(trifluoromethyl)pyridin-2-ylaminoethyl]amino-5-chloroaniline
2-[3-Chloro-5-(trifluoromethyl)pyridin-2-ylaminoethyl]amino-5-chloroaniline
Formel: C₁₄H₁₃Cl₂F₃N₄
Molecular Weight: 365,181 g/mol
Schmelzpunkt: 86…88 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00793024
CAS-Nummer: 341966-48-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR