2-Bromo-4-[(trifluoromethyl)thio]aniline 97%

Lieferant: Apollo Scientific
Achtung

PC49362-1G PC49362-250MG
APOSPC49362-1GEA 209 EUR
APOSPC49362-1G APOSPC49362-250MG
2-Bromo-4-[(trifluoromethyl)thio]aniline 97%
2-Bromo-4-[(trifluoromethyl)thio]aniline
Formel: C₇H₅BrF₃NS
Lagertemperatur: Kühlschrank
MDL: MFCD24850034
CAS-Nummer: 940077-66-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR