trans-1-Iodo-2-(heptafluoropropyl)cyclohexane
Lieferant: Apollo Scientific
Synonyme:
trans-1-(Heptafluoropropyl)-2-iodocyclohexane
PC4938-100MG
APOSPC4938-100MGEA
179
EUR
APOSPC4938-100MG
trans-1-Iodo-2-(heptafluoropropyl)cyclohexane
trans-1-Iodo-2-(heptafluoropropyl)cyclohexane
Formel:
C₉H₁₀F₇I Molecular Weight: 378,07 g/mol Siedepunkt: 48 °C/1,3mmHg Dichte: 1,792 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD01862015 CAS-Nummer: 7589-44-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...