S-(2-Amino-4-fluorophenyl) dimethylcarbamothioate 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-[(Dimethylcarbamoyl)sulphanyl]-5-fluoroaniline, S-(2-Amino-4-fluorophenyl) dimethylthiocarbamate

PC49381-1G PC49381-250MG
APOSPC49381-1GEA 557 EUR
APOSPC49381-1G APOSPC49381-250MG
S-(2-Amino-4-fluorophenyl) dimethylcarbamothioate 97%
S-(2-Amino-4-fluorophenyl) dimethylcarbamothioate
Formel: C₉H₁₁FN₂OS
Lagertemperatur: Raumtemperatur
MDL: MFCD24636943
CAS-Nummer: 1820674-43-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR