2-Bromo-3-(trifluoromethyl)benzyl bromide 95%
Lieferant: Apollo Scientific
|
Gefahr
|
PC49388-1G
PC49388-5G
PC49388-25G
APOSPC49388-1GEA
0
EUR
APOSPC49388-1G
APOSPC49388-5G
APOSPC49388-25G
2-Bromo-3-(trifluoromethyl)benzyl bromide 95%
2-Bromo-3-(trifluoromethyl)benzyl bromide
Formel:
C₈H₅Br₂F₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD13185412 CAS-Nummer: 1214372-35-6 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...