1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-1H-indole-3-carbonitrile 97%
Lieferant: Apollo Scientific
PC4940-100MG
APOSPC4940-100MGEA
271
EUR
APOSPC4940-100MG
1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-1H-indole-3-carbonitrile 97%
1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-1H-indole-3-carbonitrile
Formel:
C₁₅H₇ClF₃N₃ Molecular Weight: 321,684 g/mol Schmelzpunkt: 134…136 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00664513 CAS-Nummer: 151388-01-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...