4-Cyclopropyl-1-fluoro-2-nitrobenzene

Lieferant: Apollo Scientific
Achtung

PC49407-1G PC49407-250MG PC49407-5G
APOSPC49407-1GEA 0 EUR
APOSPC49407-1G APOSPC49407-250MG APOSPC49407-5G
4-Cyclopropyl-1-fluoro-2-nitrobenzene
4-Cyclopropyl-1-fluoro-2-nitrobenzene
Formel: C₉H₈FNO₂
Lagertemperatur: Gefrierschrank
MDL: MFCD17677330
CAS-Nummer: 769159-85-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR