1-Cyclopropyl-3-fluoro-2-nitrobenzene

Lieferant: Apollo Scientific
Achtung

PC49408-1G PC49408-250MG PC49408-5G
APOSPC49408-1GEA 0 EUR
APOSPC49408-1G APOSPC49408-250MG APOSPC49408-5G
1-Cyclopropyl-3-fluoro-2-nitrobenzene
1-Cyclopropyl-3-fluoro-2-nitrobenzene
Formel: C₉H₈FNO₂
Lagertemperatur: Gefrierschrank
MDL: MFCD31793564
CAS-Nummer: 2366994-61-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR