1-Cyclopropyl-4-fluoro-2-nitrobenzene

Lieferant: Apollo Scientific
Achtung

PC49414-1G PC49414-250MG PC49414-5G
APOSPC49414-1GEA 0 EUR
APOSPC49414-1G APOSPC49414-250MG APOSPC49414-5G
1-Cyclopropyl-4-fluoro-2-nitrobenzene
1-Cyclopropyl-4-fluoro-2-nitrobenzene
Formel: C₉H₈FNO₂
Lagertemperatur: Gefrierschrank
MDL: MFCD31793570
CAS-Nummer: 2366994-53-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR