5-Cyclopropyl-2-fluoroaniline

Lieferant: Apollo Scientific
Achtung

PC49417-1G PC49417-250MG PC49417-5G
APOSPC49417-1GEA 0 EUR
APOSPC49417-1G APOSPC49417-250MG APOSPC49417-5G
5-Cyclopropyl-2-fluoroaniline
5-Cyclopropyl-2-fluoroaniline
Formel: C₉H₁₀FN
Lagertemperatur: Gefrierschrank
MDL: MFCD24645335
CAS-Nummer: 1374298-69-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR