2-Cyclopropyl-6-fluoroaniline

Lieferant: Apollo Scientific
Achtung

PC49418-1G PC49418-250MG PC49418-5G
APOSPC49418-1GEA 0 EUR
APOSPC49418-1G APOSPC49418-250MG APOSPC49418-5G
2-Cyclopropyl-6-fluoroaniline
2-Cyclopropyl-6-fluoroaniline
Formel: C₉H₁₀FN
Lagertemperatur: Gefrierschrank
MDL: MFCD31793571
CAS-Nummer: 1701568-95-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR