2-Cyclopropyl-3-fluoroaniline

Lieferant: Apollo Scientific
Achtung

PC49419-1G PC49419-250MG PC49419-5G
APOSPC49419-1GEA 0 EUR
APOSPC49419-1G APOSPC49419-250MG APOSPC49419-5G
2-Cyclopropyl-3-fluoroaniline
2-Cyclopropyl-3-fluoroaniline
Formel: C₉H₁₀FN
Lagertemperatur: Gefrierschrank
MDL: MFCD31793572
CAS-Nummer: 2366994-64-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR