3-Cyclopropyl-5-fluoroaniline

Lieferant: Apollo Scientific
Achtung

PC49421-1G PC49421-250MG PC49421-5G
APOSPC49421-1GEA 0 EUR
APOSPC49421-1G APOSPC49421-250MG APOSPC49421-5G
3-Cyclopropyl-5-fluoroaniline
3-Cyclopropyl-5-fluoroaniline
Formel: C₉H₁₀FN
Lagertemperatur: Gefrierschrank
MDL: MFCD30210561
CAS-Nummer: 1448348-37-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR