2-Cyclopropyl-4-fluoroaniline

Lieferant: Apollo Scientific
Achtung

PC49422-1G PC49422-250MG
APOSPC49422-1GEA 0 EUR
APOSPC49422-1G APOSPC49422-250MG
2-Cyclopropyl-4-fluoroaniline
2-Cyclopropyl-4-fluoroaniline
Formel: C₉H₁₀FN
Lagertemperatur: Gefrierschrank
MDL: MFCD16778828
CAS-Nummer: 1250966-57-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR