2-Bromo-6-fluoro-4-(trifluoromethyl)benzoic acid 95%

Lieferant: Apollo Scientific
Achtung

PC49432-1G PC49432-250MG
APOSPC49432-1GEA 0 EUR
APOSPC49432-1G APOSPC49432-250MG
2-Bromo-6-fluoro-4-(trifluoromethyl)benzoic acid 95%
2-Bromo-6-fluoro-4-(trifluoromethyl)benzoic acid
Formel: C₈H₃BrF₄O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD31759997
CAS-Nummer: 1800423-60-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR