2-Bromo-4-fluoro-6-(trifluoromethyl)benzoic acid 95%

Lieferant: Apollo Scientific
Achtung

PC49433-1G PC49433-250MG
APOSPC49433-1GEA 0 EUR
APOSPC49433-1G APOSPC49433-250MG
2-Bromo-4-fluoro-6-(trifluoromethyl)benzoic acid 95%
2-Bromo-4-fluoro-6-(trifluoromethyl)benzoic acid
Formel: C₈H₃BrF₄O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD31759995
CAS-Nummer: 2091606-46-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR