2-Bromo-4-fluoro-6-(trifluoromethyl)benzoic acid 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC49433-1G
PC49433-250MG
APOSPC49433-1GEA
0
EUR
APOSPC49433-1G
APOSPC49433-250MG
2-Bromo-4-fluoro-6-(trifluoromethyl)benzoic acid 95%
2-Bromo-4-fluoro-6-(trifluoromethyl)benzoic acid
Formel:
C₈H₃BrF₄O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD31759995 CAS-Nummer: 2091606-46-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...