Ethyl 4,6-difluoro-3-methyl-1-benzofuran-2-carboxylate

Lieferant: Apollo Scientific
Achtung

PC49451-1G
APOSPC49451-1GEA 371 EUR
APOSPC49451-1G
Ethyl 4,6-difluoro-3-methyl-1-benzofuran-2-carboxylate
Ethyl 4,6-difluoro-3-methyl-1-benzofuran-2-carboxylate
Formel: C₁₂H₁₀F₂O₃
Molecular Weight: 240.203 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD32066759
CAS-Nummer: 1994412-94-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR