Methyl 2-bromo-5-chloro-4-fluorobenzoate
Lieferant: Apollo Scientific
|
Achtung
|
PC49458-1G
APOSPC49458-1GEA
1190
EUR
APOSPC49458-1G
Methyl 2-bromo-5-chloro-4-fluorobenzoate
Methyl 2-bromo-5-chloro-4-fluorobenzoate
Formel:
C₈H₅BrClFO₂ Molecular Weight: 267,48 g/mol |
MDL:
MFCD24108550 CAS-Nummer: 1502835-28-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...