4-Nitro-2-(trifluoromethyl)phenylacetic acid

Lieferant: Apollo Scientific
Achtung

PC49465-1G PC49465-250MG PC49465-5G
APOSPC49465-1GEA 226 EUR
APOSPC49465-1G APOSPC49465-250MG APOSPC49465-5G
4-Nitro-2-(trifluoromethyl)phenylacetic acid
4-Nitro-2-(trifluoromethyl)phenylacetic acid
Formel: C₉H₆F₃NO₄
Molecular Weight: 249.143 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11036389
CAS-Nummer: 199469-91-5

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR