(1R)-1-(3,5-Difluorophenyl)ethylamine 97%

Lieferant: Apollo Scientific

Synonyme: (R)-3,5-Difluoro-α-methylbenzylamine, (R)-1-(3,5-Difluorophenyl)ethanamine

PC49497-1G
APOSPC49497-1GEA 700 EUR
APOSPC49497-1G
(1R)-1-(3,5-Difluorophenyl)ethylamine 97%
(1R)-1-(3,5-Difluorophenyl)ethylamine
Formel: C₈H₉F₂N
Molecular Weight: 157,161 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06761826
CAS-Nummer: 771465-40-8
UN: 2735
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR