4-(1-Hydroxy-2,2,2-trifluoroethyl)piperidine

Lieferant: Apollo Scientific

Synonyme: 1-(Piperidin-4-yl)-2,2,2-trifluoroethan-1-ol

PC49514-1G
APOSPC49514-1GEA 1360 EUR
APOSPC49514-1G
4-(1-Hydroxy-2,2,2-trifluoroethyl)piperidine
4-(1-Hydroxy-2,2,2-trifluoroethyl)piperidine
Formel: C₇H₁₂F₃NO
Molecular Weight: 183,171 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17977194
CAS-Nummer: 743402-57-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR