4-(Difluoromethyl)-1,2-difluorobenzene 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC49547-1G
PC49547-250MG
APOSPC49547-1GEA
174
EUR
APOSPC49547-1G
APOSPC49547-250MG
4-(Difluoromethyl)-1,2-difluorobenzene 97%
4-(Difluoromethyl)-1,2-difluorobenzene
Formel:
C₇H₄F₄ Siedepunkt: 62…64 °C/62mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD14698573 CAS-Nummer: 1214379-64-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...