2-(Difluoromethyl)-1,3-difluorobenzene

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,6-Difluorobenzal fluoride

PC49548-1G
APOSPC49548-1GEA 139 EUR
APOSPC49548-1G
2-(Difluoromethyl)-1,3-difluorobenzene
2-(Difluoromethyl)-1,3-difluorobenzene
Formel: C₇H₄F₄
Molecular Weight: 164,1 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14698572
CAS-Nummer: 91624-89-4
UN: 1993
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR